Läs mer
STEMCELL Technologies ALDEFLUOR™ Assay Buffer
Produktkod. 17167711
Encompass_Preferred
Change view
Kvantitet:
25 ml
55 ml
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17167711 55 ml
17157711 25 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17167711 Leverantör STEMCELL Technologies Leverantörsnummer 01702
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Additional assay buffer for use with the ALDEFLUOR™ kit
  • ALDEFLUOR™ Assay Buffer can be used when counterstaining ALDEFLUOR™-reacted cells with immunofluorescently-labeled antibodies
  • The buffer is necessary to maintain ALDEFLUOR™ reaction product inside the cell, allowing detection of the ALDH-bright population
  • While there is sufficient ALDEFLUOR™-buffer contained within the ALDEFLUOR™ kit, additional buffer may be required for certain immunolabeling procedures
Specifikationer
För användning med (applikation) Cancer Research; Neuroscience; Stem Cell Biology
Kvantitet 55 ml