missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Aldehyde Dehydrogenase 1-A1/ALDH1A1 Antibody (M562), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23187
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Aldehyde Dehydrogenase 1-A1/ALDH1A1 Monoclonal antibody specifically detects Aldehyde Dehydrogenase 1-A1/ALDH1A1 in Human samples. It is validated for Western Blot, Immunofluorescence
Specifikationer
| Aldehyde Dehydrogenase 1-A1/ALDH1A1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, 1 mg/ml BSA and 50% glycerol | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunofluorescence | |
| M562 | |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100 | |
| acetaldehyde dehydrogenase 1, ALDC, aldehyde dehydrogenase 1 family, member A1, aldehyde dehydrogenase 1, soluble, Aldehyde dehydrogenase family 1 member A1, Aldehyde dehydrogenase, cytosolic, aldehyde dehydrogenase, liver cytosolic, ALDH class 1, ALDH1, ALDH11, ALDH-E1, ALHDII, EC 1.2.1, EC 1.2.1.36, MGC2318, PUMB1, RALDH 1, RalDH1, retinal dehydrogenase 1, retinaldehyde dehydrogenase 1 | |
| Clone M562 was generated from a recombinant protein corresponding to amino acids in the N-terminal region from human Aldehyde Dehydrogenase 1-A1/ALDH1A1. [UniProt# P00352] | |
| 0.1 mL | |
| Cancer, Diabetes Research, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience, Neurotransmission, Stem Cell Markers | |
| 216 | |
| Store at -20°C. | |
| IgG2a |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering