missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ ALDH3B1 Polyclonal Antibody

Description
ALDH3B1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
The aldehyde dehydrogenases are a family of isozymes that may play a major role in the detoxification of aldehydes generated by alcohol metabolism and lipid peroxidation. This particular gene spans about 20 kb of genomic DNA and is composed of 9 coding exons. The gene is highly expressed in kidney and lung. The functional significance of this gene as well as the cellular localization of its product are presently unknown. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
| Antigen | ALDH3B1 |
| Användningsområden | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Klassificering | Polyclonal |
| Koncentration | 0.7 mg/mL |
| Konjugera | Unconjugated |
| Formulering | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gen | ALDH3B1 |
| Genaccessionsnr. | P43353, Q5XI42, Q80VQ0 |
| Gene Alias | 1700001N19Rik; aldehyde dehydrogenase 3 family member B1; aldehyde dehydrogenase 3 family, member B1; aldehyde dehydrogenase 3B1; aldehyde dehydrogenase 7; Aldehyde dehydrogenase family 3 member B1; Aldh3b1; ALDH4; ALDH7; FLJ26433; FLJ34710 |
| Gensymboler | ALDH3B1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?