missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALDH9A1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26621-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
ALDH9A1 belongs to the aldehyde dehydrogenase family of proteins. The protein has a high activity for oxidation of gamma-aminobutyraldehyde and other amino aldehydes. The enzyme catalyzes the dehydrogenation of gamma-aminobutyraldehyde to gamma-aminobutyric acid (GABA). This isozyme is a tetramer of identical 54-kD subunits.Specifications
ALDH9A1 | |
Polyclonal | |
Unconjugated | |
ALDH9A1 | |
ALDH4, ALDH7, ALDH9, ALDH9A1, E3, TMABADH | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
223 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P49189 | |
ALDH9A1 | |
ALDH9A1 Fusion Protein Ag24312 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |