Beskrivning
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
- Expert Technical Support
Specifikationer
Specifikationer
| Kemiskt namn eller material | Aldicarb-sulfoxide |
| Förpackning | Glass Bottle |
| CAS | 1646-87-3 |
| Färg | Colorless |
| Smältpunkt | 108°C to 110°C |
| Synonym | 2-Methyl-2-(methylsulfinyl)propanal O-[(Methylamino)carbonyl]oxime |
| Industry Type | Environmental Analysis |
| Molekylvikt (g/mol) | 206.26 |
| Kvantitet | 100 mg |
| Kvalitet | Analytical Standard |