Learn More
Abnova™ ALDOA (Human) Recombinant Protein
Human ALDOA full-length ORF ( AAH10660.1, 1 a.a. - 364 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00000226-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MPYQYPALTPEQKKELSDIAHRIVAPGKGILAADESTGSIAKRLQSIGTENTEENRRFYRQLLLTADDRVNPCIGGVILFHETLYQKADDGRPFPQVIKSKGGVVGIKVDKGVVPLAGTNGETTTQGLDGLSERCAQYKKDGADFAKWRCVLKIGEHTPSALAIMENANVLARYASICQQNGIVPIVEPEILPDGDHDLKRCQYVTEKVLAAVYKALSDHHIYLEGTLLKPNMVTPGHACTQKFSHEEIAMATVTALRRTVPPAVTGITFLSGGQSEEEASINLNAINKCPLLKPWALTFSYGRALQASALKAWGGKKENLKAAQEEYVKRALANSLACQGKYTPSGQAGAAASESLFVSNHAY
Specifications
AAH10660.1 | |
aldolase A, fructose-bisphosphate | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ALDOA | |
GST |
226 | |
Wheat germ expression system | |
10 μg | |
ALDA, MGC10942, MGC17716, MGC17767 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |