Läs mer
Aldosterone 3-(O-Carboxymethyl)oxime (>80%), TRC
Produktkod. 30341803
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30341803 10 mg
1 options
Produktkod. 30341803 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA66593010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Aldosterone 3-(O-Carboxymethyl)oxime
CAS 55482-95-6
Molekylformel C23 H31 N O7
Kvantitet 10 mg
Synonym [[[(11β)-11,21-Dihydroxy-18,20-dioxopregn-4-en-3-ylidene]amino]oxy]acetic Acid, Pregnane Acetic Acid Derivative, 4-Pregnen-11β,21-diol-3,18,20-trione 3-O-Carboxymethyloxime, 2-((((8S,9S,10R,11S,13R,14S,17S,E)-13-Formyl-11-hydroxy-17-(2-hydroxyacetyl)-10-methyl-1,2,6,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-3H-cyclopenta[a]phenanthren-3-ylidene)amino)oxy)acetic Acid
InChI formel InChI=1S/C23H31NO7/c1-22-7-6-14(24-31-11-20(29)30)8-13(22)2-3-15-16-4-5-17(19(28)10-25)23(16,12-26)9-18(27)21(15)22/h8,12,15-18,21,25,27H,2-7,9-11H2,1H3,(H,29,30)/b24-14+/t15-,16-,17+,18-,21+,22-,23+/m0/s1
LEDER C[C@]12CC\C(=N/OCC(=O)O)\C=C1CC[C@H]3[C@@H]4CC[C@H](C(=O)CO)[C@]4(C[C@H](O)[C@H]23)C=O
IUPAC-namn 2-[(E)-[(8S,9S,10R,11S,13R,14S,17S)-13-formyl-11-hydroxy-17-(2-hydroxyacetyl)-10-methyl-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-ylidene]amino]oxyacetic acid
Molekylvikt (g/mol) 433.49
Formel vikt 433.2101