missing translation for 'onlineSavingsMsg'
Läs mer

Alectinib M4 metabolite, TRC

Produktkod. 30138548
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
2.5 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30138548 10 mg
30134769 5 mg
30011566 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138548 Supplier Toronto Research Chemicals Supplier No. TRCA52115010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1256585-04-2
Molekylformel C28H32N4O2
Molekylvikt (g/mol) 456.58
Synonym 9-Ethyl-6,11-dihydro-8-[4-[(2-hydroxyethyl)amino]-1-piperidinyl]-6,6-dimethyl-11-oxo-5H-benzo[b]carbazole-3-carbonitrile
IUPAC-namn 9-ethyl-8-[4-(2-hydroxyethylamino)piperidin-1-yl]-6,6-dimethyl-11-oxo-5H-benzo[b]carbazole-3-carbonitrile
LEDER OCCNC1CCN(C2=CC(C(C)(C)C(C(C=CC(C#N)=C3)=C3N4)=C4C5=O)=C5C=C2CC)CC1

Specifications

Kemiskt namn eller material Alectinib M4 metabolite
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C28H32N4O2/c1-4-18-14-21-22(15-24(18)32-10-7-19(8-11-32)30-9-12-33)28(2,3)27-25(26(21)34)20-6-5-17(16-29)13-23(20)31-27/h5-6,13-15,19,30-31,33H,4,7-12H2,1-3H3
Procent renhet >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.