Learn More
Thermo Scientific Alfa Aesar Al-23 Crucible, Conical,Ht (mm), 55,Top OD (mm), 65,Top ID (mm), 59,Base OD (mm), 33,Volume (ml), 80
Brand: Thermo Scientific Alfa Aesar 033047.KF
Al-23 Crucible, Conical,Ht (mm), 55,Top OD (mm), 65,Top ID (mm), 59,Base OD (mm), 33,Volume (ml), 80
Additional Details : Weight : 0.01000kg