Learn More
Thermo Scientific Alfa Aesar Al-23 Crucible, Conical,Ht (mm), 38,Top OD (mm), 30,Top ID (mm), 26,Base OD (mm), 15,Volume (ml), 15
Brand: Thermo Scientific Alfa Aesar 033011.KF
Al-23 Crucible, Conical,Ht (mm), 38,Top OD (mm), 30,Top ID (mm), 26,Base OD (mm), 15,Volume (ml), 15
Additional Details : Weight : 0.01000kg