Learn More
Thermo Scientific Alfa Aesar Al-25 Crucible, Cylindrical, Flat Base,Height x OD x ID (mm), 200 x 120 x 100,Vol (ml), 1500 1piece
Brand: Thermo Scientific Alfa Aesar 032968.KF
Al-25 Crucible, Cylindrical, Flat Base,Height x OD x ID (mm), 200 x 120 x 100,Vol (ml), 1500 1piece
Additional Details : Weight : 0.01000kg