Learn More
Alfa Aesar™ Glass Ceramic Rectangular Plate, L x W: 100 x 150mm
Brand:  Alfa Aesar 045720.KS
Additional Details : Weight : 0.01000kg
Product Code. 15464015
  • 2439.30 SEK / Each
Estimated Shipment date
from Supplier 06-02-2018
Add to basket
Specifications
Specifications
4 x 6 in. (100 x 150mm)
Rectangular
Glass Ceramic
150mm
1 Ea.
100mm
Glass Ceramic Rectangular Plate