Läs mer
Algestone Acetonide, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Algestone Acetonide |
| CAS | 4968-09-6 |
| Molekylformel | C24H34O4 |
| Kvantitet | 25 mg |
| Synonym | 16alpha,17-dihydroxy-Pregn-4-ene-3,20-dione cyclic acetal with acetone (7CI,8CI), Progesterone, 16alpha,17-dihydroxy-, cyclic acetal with acetone (6CI), (16alpha)-16,17-[(1-Methylethylidene)bis(oxy)]pregn-4-ene-3,20-dione, 16alpha,17-(Isopropylidenedioxy)progesterone, 16alpha,17-Dihydroxypregn-4-ene-3,20-dione cyclic acetal with acetone, 16alpha,17alpha-Isopropylidenedioxyprogesterone, Alphasone acetonide, NSC 26104, Progesterone 16,17-acetonide, W 3395, (16alpha)-16,17-[(1-Methylethylidene)bis(oxy)]-pregn-4-ene-3,20-dione |
| LEDER | CC(=O)[C@@]12OC(C)(C)O[C@@H]1C[C@H]3[C@@H]4CCC5=CC(=O)CC[C@]5(C)[C@H]4CC[C@]23C |
| Molekylvikt (g/mol) | 386.52 |
| Formel vikt | 386.2457 |