Läs mer
Algestone Acetophenide, TRC
Produktkod. 17913075
Change view
Kvantitet:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17913075 5 g
17960655 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17913075 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA5320405G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 24356-94-3
Molekylformel C29 H36 O4
Synonym Pregn-4-ene-3,20-dione, 16,17-[[(1R)-1-phenylethylidene]bis(oxy)]-, (16α)-∣Pregn-4-ene-3,20-dione, 16,17-[(1-phenylethylidene)bis(oxy)]-, [16α(R)]-∣Pregn-4-ene-3,20-dione, 16α,17-dihydroxy-, cyclic acetal with acetophenone, (R)- (8CI)∣2H-Naphth[1',2':6,7]indeno[1,2-d][1,3]dioxole, pregn-4-ene-3,20-dione deriv.∣Algestone acetophenide∣Alphasone acetophenide∣Deladroxone∣Dihydroxyprogesterone acetophenide∣Droxone∣Neolutin Depositum∣P-DHP∣ZK 9349∣2H-Naphth[1',2':6,7]indeno[1,2-d][1,3]dioxole, pregn-4-ene-3,20-dione deriv.∣Algestone acetophenide∣Alphasone acetophenide∣Deladroxone∣Dihydroxyprogesterone acetophenide∣Droxone∣Neolutin Depositum∣P-DHP∣ZK 9349∣Pregn-4-ene-3,20-dione, 16,17-[[(1R)-1-phenylethylidene]bis(oxy)]-, (16alpha)-∣Pregn-4-ene-3,20-dione, 16,17-[(1-phenylethylidene)bis(oxy)]-, [16alpha(R)]-∣Pregn-4-ene-3,20-dione, 16alpha,17-dihydroxy-, cyclic acetal with acetophenone, (R)- (8CI)
Specifikationer
Kemiskt namn eller material Algestone Acetophenide
Kvantitet 5 g
Frakt skick Room Temperature
Formel vikt 448.2614
Molekylvikt (g/mol) 448.59, 448.60
InChI-nyckel AHBKIEXBQNRDNL-JRLVXNONSA-N
LEDER CC(=O)[C@@]12OC(C)(O[C@@H]1C[C@H]1C3CCC4=CC(=O)CCC4(C)[C@H]3CCC21C)C1=CC=CC=C1
IUPAC-namn (2S,4R,8S,12S)-8-acetyl-6,9,13-trimethyl-6-phenyl-5,7-dioxapentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icos-17-en-16-one