Läs mer
Algestone, TRC
Produktkod. 30136270
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136270 500 mg
30009574 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136270 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA532030500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 595-77-7
Molekylformel C21 H30 O4
Molekylvikt (g/mol) 346.46
Synonym (16α)-16,17-Dihydroxy-pregn-4-ene-3,20-dione, Pregn-4-ene-3,20-dione, 16α,17-dihydroxy- (8CI), 16α,17-Dihydroxyprogesterone, 16α,17α-Dihydroxypregn-4-ene-3,20-dione, 16α,17α-Dihydroxyprogesterone, 4-Pregnen-16α,17α-diol-3,20-dione, Alfasone, Algestone, Alphasone
IUPAC-namn (8R,9S,10R,13S,14S,16R,17S)-17-acetyl-16,17-dihydroxy-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one
LEDER CC(=O)[C@@]1(O)[C@H](O)C[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
Specifikationer
Kemiskt namn eller material Algestone
Analyt- eller komponentnamn Algestone
Anteckningar om renhetsgrad HPLC
Frakt skick Room Temperature
InChI formel InChI=1S/C21H30O4/c1-12(22)21(25)18(24)11-17-15-5-4-13-10-14(23)6-8-19(13,2)16(15)7-9-20(17,21)3/h10,15-18,24-25H,4-9,11H2,1-3H3/t15-,16+,17+,18-,19+,20+,21-/m1/s1
Lösningstyp Neat
Industry Type Pharmaceutical
Kvantitet 500 mg
Formel vikt 346.2144
Procent renhet >95%