Läs mer
Aliskiren Hydrochloride, TRC
Produktkod. 30322344
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30322344 10 mg
1 options
Produktkod. 30322344 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA53600010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Aliskiren
CAS 173399-03-6
Molekylformel C30 H53 N3 O6 . Cl H
Kvantitet 10 mg
Synonym (alphaS,deltaS,gammaS,zetaS)-delta-Amino-N-(3-amino-2,2-dimethyl-3-oxopropyl)-gamma-hydroxy-4-methoxy-3-(3-methoxypropoxy)-alpha,zeta-bis(1-methylethyl)benzeneoctanamide Hydrochloride (1:1), Benzeneoctanamide, delta-amino-N-(3-amino-2,2-dimethyl-3-oxopropyl)-gamma-hydroxy-4-methoxy-3-(3-methoxypropoxy)-alpha,zeta-bis(1-methylethyl)-, monohydrochloride, (alphaS,gammaS,deltaS,zetaS)- (9CI), Benzeneoctanamide, delta-amino-N-(3-amino-2,2-dimethyl-3-oxopropyl)-gamma-hydroxy-4-methoxy-3-(3-methoxypropoxy)-alpha,zeta-bis(1-methylethyl)-, monohydrochloride, [alphaS-(alphaR*,gammaR*,deltaR*,zetaR*)]-, Aliskiren hydrochloride
InChI formel InChI=1S/C30H53N3O6.ClH/c1-19(2)22(14-21-10-11-26(38-8)27(15-21)39-13-9-12-37-7)16-24(31)25(34)17-23(20(3)4)28(35)33-18-30(5,6)29(32)36;/h10-11,15,19-20,22-25,34H,9,12-14,16-18,31H2,1-8H3,(H2,32,36)(H,33,35);1H/t22-,23-,24-,25-;/m0./s1
LEDER Cl.COCCCOc1cc(C[C@@H](C[C@H](N)[C@@H](O)C[C@@H](C(C)C)C(=O)NCC(C)(C)C(=O)N)C(C)C)ccc1OC
IUPAC-namn (2S,4S,5S,7S)-5-amino-N-(3-amino-2,2-dimethyl-3-oxopropyl)-4-hydroxy-7-[[4-methoxy-3-(3-methoxypropoxy)phenyl]methyl]-8-methyl-2-propan-2-ylnonanamide;hydrochloride
Molekylvikt (g/mol) 588.22
Formel vikt 587.3701