Learn More
Alkaline Phosphatase, Intestinal Antibody (AP-7/6/7), Biotin, R&D Systems™
Product Code. 18607917 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity Unit Size
18607917 0.1 mL 0.1mL
1 options
Product Code. 18607917 Supplier Novus Biologicals Supplier No. NBP250102B
 Restricted Item
Beräknad leverans: 14-12-2026
A Novus product, part of R&D Systems. Mouse Monoclonal Antibody
Alkaline Phosphatase, Intestinal Monoclonal antibody specifically detects Alkaline Phosphatase, Intestinal in Bovine samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Antigen Alkaline Phosphatase, Intestinal
Applications Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone AP-7/6/7
Conjugate Biotin
Formulation PBS
Gene Alias alkaline phosphatase, intestinal, alkaline phosphomonoesterase, EC 3.1.3.1, glycerophosphatase, IAP, Intestinal alkaline phosphatase, intestinal-type alkaline phosphatase, Kasahara isozyme
Host Species Mouse
Immunogen Calf intestinal alkaline phosphatase
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Ovarian Carcinoma Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 248
Target Species Bovine
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line Novus