Läs mer
Alkaline Phosphatase, MP Biomedicals™
Produktkod. 11481431 Handla allt MP Biomedicals Produkter
Change view
Kvantitet:
5 mg
10 mg
25 mg
Förpackningsstorlek:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11481431 5 mg 5mg
11491431 10 mg 10mg
11401441 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 11481431 Leverantör MP Biomedicals Leverantörsnummer 0215934705
Specifikationer
CAS 9001-78-9
Innehåll och lagring Store at 0°C.
Formatera Solid
Molekylvikt (g/mol) 496.569
Produkttyp Alkaline Phosphatase