missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALKBH4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19882-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Dioxygenase that mediates demethylation of actin monomethylated at ′Lys-84′ (K84me1), thereby acting as a regulator of actomyosin-processes. Demethylation of actin K84me1 is required for maintaining actomyosin dynamics supporting normal cleavage furrow ingression during cytokinesis and cell migration. May be involved in transcription regulation.Specifications
ALKBH4 | |
Polyclonal | |
Unconjugated | |
ALKBH4 | |
ABH4, ALKBH4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
288587, 54784, 72041 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9D8F1, Q9NXW9 | |
Alkbh4 | |
ALKBH4 Fusion Protein Ag8041 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |