Läs mer
Alkylated Diphenylamines, TRC
Produktkod. 30005710
Change view
Kvantitet:
250 mg
1 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30005710 25 g
30010865 250 mg
30004426 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30005710 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA57510025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 68921-45-9
Molekylformel C20H27N
Molekylvikt (g/mol) 281.44
Synonym n-phenyl-4-(2,4,4-trimethylpentan-2-yl)aniline Benzenamine, N-phenyl-ar-(1,1,3,3-tetramethylbutyl)-4-tert-octyldiphenylamine
IUPAC-namn N-phenyl-4-(2,4,4-trimethylpentan-2-yl)aniline
LEDER CC(C1=CC=C(C=C1)NC2=CC=CC=C2)(C)CC(C)(C)C
Specifikationer
Kemiskt namn eller material Alkylated Diphenylamines
Kvantitet 25 g
InChI formel InChI=1S/C20H27N/c1-19(2,3)15-20(4,5)16-11-13-18(14-12-16)21-17-9-7-6-8-10-17/h6-14,21H,15H2,1-5H3
Formel vikt 281.21