missing translation for 'onlineSavingsMsg'
Läs mer

all-trans 5,8-Epoxy Retinoic Acid (Mixture of Diastereomers), TRC

Produktkod. 30373073
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30373073 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. View return policy
Product Code. 30373073 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCE5898051MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material all-trans 5,8-Epoxy Retinoic acid (mixture of diastereomers)
CAS 3012-76-8
Molekylformel C20 H28 O3
Kvantitet 1 mg
Synonym (2E,4E,6E)-7-(2,4,5,6,7,7a-Hexahydro-4,4,7a-trimethyl-2-benzofuranyl)-3-methyl-2,4,6-octatrienoic Acid, 5,8-Epoxy-5,8-dihydroretinoic Acid, 5,8-Monoepoxyretinoic Acid
InChI formel InChI=1S/C20H28O3/c1-14(12-18(21)22)8-6-9-15(2)16-13-17-19(3,4)10-7-11-20(17,5)23-16/h6,8-9,12-13,16H,7,10-11H2,1-5H3,(H,21,22)/b8-6+,14-12+,15-9+
LEDER C\C(=C/C(=O)O)\C=C\C=C(/C)\C1OC2(C)CCCC(C)(C)C2=C1
IUPAC-namn (2E,4E,6E)-7-(4,4,7a-trimethyl-2,5,6,7-tetrahydrobenzofuran-2-yl)-3-methyl-octa-2,4,6-trienoic acid
Molekylvikt (g/mol) 316.43
Formel vikt 316.2038
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.