Läs mer
all-trans-Anhydro Retinol (90%), TRC
Produktkod. 30343021
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30343021 50 mg
1 options
Produktkod. 30343021 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA65450050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material all-trans-Anhydro Retinol
CAS 1224-78-8
Molekylformel C20 H28
Kvantitet 50 mg
Synonym Cyclohexene, 6-[(2E,4E,6E)-3,7-dimethyl-2,4,6,8-nonatetraen-1-ylidene]-1,5,5-trimethyl-, (6E)-, 1,3,5,7-Nonatetraene, 3,7-dimethyl-9-(2,6,6-trimethyl-2-cyclohexen-1-ylidene)-, (all-E)- (8CI), Cyclohexene, 6-(3,7-dimethyl-2,4,6,8-nonatetraenylidene)-1,5,5-trimethyl-, (all-E)-, Cyclohexene, 6-[(2E,4E,6E)-3,7-dimethyl-2,4,6,8-nonatetraenylidene]-1,5,5-trimethyl-, (6E)- (9CI), (6E)-6-[(2E,4E,6E)-3,7-Dimethyl-2,4,6,8-nonatetraen-1-ylidene]-1,5,5-trimethylcyclohexene, Anhydroretinol, Anhydrovitamin A, Anhydrovitamin A1, R368, R368 (vitamin analog), Retinol, anhydro-, Vitamin A1, anhydro-, all-trans-Anhydroretinol, trans-Anhydrovitamin A
InChI formel InChI=1S/C20H28/c1-7-16(2)10-8-11-17(3)13-14-19-18(4)12-9-15-20(19,5)6/h7-8,10-14H,1,9,15H2,2-6H3/b11-8+,16-10+,17-13+,19-14-
LEDER C\C(=C/C=C/C(=C/C=C\1/C(=CCCC1(C)C)C)/C)\C=C
IUPAC-namn (6E)-6-[(2E,4E,6E)-3,7-dimethylnona-2,4,6,8-tetraenylidene]-1,5,5-trimethylcyclohexene
Molekylvikt (g/mol) 268.44
Formel vikt 268.2191