Läs mer
Allantoic Acid, TRC
Produktkod. 30133401
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133401 500 mg
17981885 50 mg
17983165 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133401 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA541500500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 99-16-1
Molekylformel C4 H8 N4 O4
Molekylvikt (g/mol) 176.13
Synonym 2,2-Bis[(aminocarbonyl)amino]acetic Acid, Diureido-acetic Acid;
IUPAC-namn 2,2-bis(carbamoylamino)acetic acid
LEDER NC(=O)NC(NC(=O)N)C(=O)O
Specifikationer
Kemiskt namn eller material Allantoic Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 500 mg
InChI formel InChI=1S/C4H8N4O4/c5-3(11)7-1(2(9)10)8-4(6)12/h1H,(H,9,10)(H3,5,7,11)(H3,6,8,12)
Formel vikt 176.0546
Procent renhet >95%