Läs mer
Allantoin-13C2,15N4, TRC
Produktkod. 17998704
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17998704 1 mg
17988894 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17998704 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA5405021MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1219402-51-3
Molekylformel C213C2H615N4O3
Molekylvikt (g/mol) 164.07
Synonym 5-Ureidohydantoin-13C2,15N4, (2,5-Dioxo-4-imidazolidinyl)urea-13C2,15N4, Allantol-13C2,15N4, Cordianine-13C2,15N4, Glyoxyldiureid-13C2,15N4, NSC 7606-13C2,15N4, Psoralon-13C2,15N4, Sebical-13C2,15N4, Septalan-13C2,15N4
IUPAC-namn (2,5-dioxo-(213C,1,3-15N2)1,3-diazolidin-4-yl)(13C)urea
LEDER O=[13C]([15NH]1)[15NH]C([15NH][13C]([15NH2])=O)C1=O
Specifikationer
Kemiskt namn eller material Allantoin-13C2,15N4
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C4H6N4O3/c5-3(10)6-1-2(9)8-4(11)7-1/h1H,(H3,5,6,10)(H2,7,8,9,11)/i3+1,4+1,5+1,6+1,7+1,8+1
Procent renhet >95%