Learn More
Alfa Aesar™ Allatostatin II
For cellular and molecular biology applications
Brand:  Alfa Aesar™ J66779.#0
Additional Details : Weight : 0.00005kg
 View more versions of this product
Product Code. 15448269
  • 2545.35 SEK / 5mg
Estimated Shipment date
from Supplier 12-06-2018
Add to basket
Specifications
Specifications
1067.19
5mg
Gly-Asp-Gly-Arg-Leu-Tyr-Ala-Phe-Gly-Leu-NH2
-30° to -10°C
HPLC
Solid
RUO
Allatostatin II