Läs mer
Allethrin, TRC
Produktkod. 30040781
Change view
Kvantitet:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30040781 500 mg
30012552 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30040781 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA542200500MG
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 584-79-2
Molekylformel C19 H26 O3
Molekylvikt (g/mol) 302.41
Synonym Chrysanthemummonocarboxylic acid, 3-allyl-2-methyl-4-oxo-2-cyclopenten-1-yl ester (6CI), Cyclopropanecarboxylic acid, 2,2-dimethyl-3-(2-methyl-1-propenyl)-, 2-methyl-4-oxo-3-(2-propenyl)-2-cyclopenten-1-yl ester (9CI), Cyclopropanecarboxylic acid, 2,2-dimethyl-3-(2-methylpropenyl)-, ester with 2-allyl-4-hydroxy-3-methyl-2-cyclopenten-1-one (8CI), 2-Cyclopenten-1-one, 2-allyl-4-hydroxy-3-methyl-, 2,2-dimethyl-3-(2-methylpropenyl)cyclopropanecarboxylate (8CI), (±)-3-Allyl-4-keto-2-methylcyclopent-2-enyl (±)-cis,trans-chrysanthemummonocarboxylate, (±)-Allerethonyl (±)-cis,trans-chrysanthemate, 2,2-Dimethyl-3-(2-methyl-1-propenyl)cyclopropanecarboxylic acid, ester with 2-allyl-4-hydroxy-3-methyl-2-cyclopenten-1-one, 2-Allyl-4-hydroxy-3-methyl-2-cyclopenten-1-one ester of 2,2-dimethyl-3-(2-methylpropenyl)cyclopropane carboxylic acid, 2-Methyl-3-(2-propenyl)-4-oxo-2-cyclopentenyl 2,2-dimethyl-3-(2-methyl-1-propenyl)cyclopropanecarboxylate, 3-(2-Propenyl)-2-methyl-4-oxo-2-cyclopentenyl 3,3-dimethyl-2-(2-methyl-1-propenyl)cyclopropanecarboxylate, 3-Allyl-4-keto-2-methylcyclopentenyl chrysanthemummonocarboxylate, 3-Allyl-4-methyl-2-oxo-3-cyclopenten-1-yl ester of 2,2-dimethyl-3-(2-methylpropenyl)cyclopropanecarboxylic acid, Allethrin, Allethrine, Allyl cinerin I, Depallethrin, Depallethrine, EBT, ENT 17510, Exthrin, Matox, NSC 11782, OMS 3045, Pynamin, Pyresyn, RU 27436, Wasp Stopper CF, dl, d-T80 allethrin, dl-2-Allyl-4-hydroxy-3-methyl-2-cyclopenten-1-one ester with dl-chrysanthemummonocarboxylic acid, dl-Allethrin, trans-Allethrin
IUPAC-namn (2-methyl-4-oxo-3-prop-2-enylcyclopent-2-en-1-yl) 2,2-dimethyl-3-(2-methylprop-1-enyl)cyclopropane-1-carboxylate
LEDER CC(=CC1C(C(=O)OC2CC(=O)C(=C2C)CC=C)C1(C)C)C
Specifikationer
Kemiskt namn eller material Allethrin
Kvantitet 500 mg
InChI formel InChI=1S/C19H26O3/c1-7-8-13-12(4)16(10-15(13)20)22-18(21)17-14(9-11(2)3)19(17,5)6/h7,9,14,16-17H,1,8,10H2,2-6H3
Formel vikt 302.1882