Learn More
Witeg™ Allihn Bulb Condensers
Product Code. 11833562
Change view
Click to view available options
Standard Taper Joint No:
19/26
24/29
29/32
Length (Metric) Jacket:
250 mm
400 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No Length (Metric) Jacket Unit Size
11833562 19/26 250 mm Each
11843562 24/29 250 mm Each
11863562 29/32 250 mm Each
11873562 29/32 400 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11833562 Supplier Witeg™ Supplier No. 1 722 250
  • ONLINE EXCLUSIVE PRICE
    1615.00 SEK
    867.37 SEK / Paket med 1
    Save 747.63 SEK
    46% Av
Beräknad leverans: 16-12-2026
Use the Witeg™ Allihn Bulb Condenser, which consists of a long glass tube with a water jacket. A series of bulbs on the tube increases the surface area.
  • Various jacket lengths
Specifications
Diameter (Metric) Inner Tubing 6.4 mm
Diameter (Metric) Outer Tubulation 11 mm
Length (Metric) Jacket 250 mm
Quantity 1 Pack
Standard Taper Joint No 19/26
Type Allihn Bulb Condenser