Learn More
Allihn Condenser with Ground Cone and Socket
Product Code. 11381845
Change view
Click to view available options
Standard Taper Joint No:
24/29
29/32
Length (Metric) Jacket:
545 mm
550 mm
Unit Size:
Each
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No Length (Metric) Jacket Unit Size
11381845 24/29 545 mm Each
15507790 29/32 550 mm Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11381845 Supplier Quickfit Supplier No. CX7/33/SC
  • ONLINE EXCLUSIVE PRICE
    2565.00 SEK
    2115.88 SEK / Paket med 1
    Save 449.12 SEK
    17% Av
Beräknad leverans: 10-03-2027
Ideal for reflux applications.
  • Inner bulb sections provide increased cooling surface
  • With detachable plastic screwthread connectors for easy and safe attachment of flexible tubing
Specifications
For Use With (Application) Ideal for reflux work
Length (Metric) Jacket 545 mm
Length (Metric) 545 mm
Standard Taper Joint No 24/29
Material Borosilicate Glass
Type Allihn Condenser