missing translation for 'onlineSavingsMsg'
Läs mer

Allyl 4-Aminopiperdine-1-carboxylate, TRC

Produktkod. 30324202
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30324202 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30324202 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA5495801G

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material Allyl 4-Aminopiperdine-1-carboxylate
CAS 358969-71-8
Molekylformel C9 H16 N2 O2
Kvantitet 1 g
Synonym 4-amino-1-N-alloc-piperidine, 4-Amino-piperidine-1-carboxylic Acid Allyl Ester, 4-Amino-2-propen-1-yl Ester 1-Piperidinecarboxylic Acid
InChI formel InChI=1S/C9H16N2O2/c1-2-7-13-9(12)11-5-3-8(10)4-6-11/h2,8H,1,3-7,10H2
LEDER NC1CCN(CC1)C(=O)OCC=C
IUPAC-namn prop-2-enyl 4-aminopiperidine-1-carboxylate
Molekylvikt (g/mol) 184.24
Formel vikt 184.1212
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.