Läs mer
Allyl b-D-Glucopyranoside, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Allyl b-D-Glucopyranoside |
| CAS | 34384-79-7 |
| Molekylformel | C9 H16 O6 |
| Kvantitet | 500 mg |
| Synonym | Glucopyranoside, allyl, β-D- (8CI), β-D-Glucopyranoside, 2-propenyl (9CI), 2-Propen-1-yl β-D-glucopyranoside, Allyl β-D-glucopyranoside |
| InChI formel | InChI=1S/C9H16O6/c1-2-3-14-9-8(13)7(12)6(11)5(4-10)15-9/h2,5-13H,1,3-4H2/t5-,6-,7+,8-,9-/m1/s1 |
| LEDER | OC[C@H]1O[C@@H](OCC=C)[C@H](O)[C@@H](O)[C@@H]1O |
| IUPAC-namn | (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-prop-2-enoxyoxane-3,4,5-triol |
| Molekylvikt (g/mol) | 220.22 |
| Formel vikt | 220.095 |