Läs mer
Allyl Ester of Atorvastatin Cyclic (Fluorophenyl) Impurity, TRC
Produktkod. 30348083
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30348083 1 mg
1 options
Produktkod. 30348083 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA7918301MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Allyl Ester of Atorvastatin Cyclic (Fluorophenyl) Impurity
CAS 1316643-57-8
Molekylformel C36H39FN2O7
Kvantitet 1 mg
Synonym 1beta-(4-Fluorophenyl)hexahydro-beta,7-dihydroxy-7-(1-methylethyl)-1alpha-phenyl-7alpha-[(phenylamino)carbonyl]-3H-oxireno[3,4]pyrrolo[2,1-b][1,3]oxazine-3-butanoic Acid Allyl Ester, Allyl-ATV-cycloFP
InChI formel InChI=1S/C36H39FN2O7/c1-4-21-44-31(41)23-29(40)22-30-19-20-39-35(43,24(2)3)34(32(42)38-28-13-9-6-10-14-28)33(46-34,25-11-7-5-8-12-25)36(39,45-30)26-15-17-27(37)18-16-26/h4-18,24,29-30,40,43H,1,19-23H2,2-3H3,(H,38,42)
LEDER CC(C)C1(O)N2CCC(CC(O)CC(=O)OCC=C)OC2(c3ccc(F)cc3)C4(OC14C(=O)Nc5ccccc5)c6ccccc6
IUPAC-namn prop-2-enyl 4-[1b-(4-fluorophenyl)-7-hydroxy-1a-phenyl-7a-(phenylcarbamoyl)-7-propan-2-yl-4,5-dihydro-3H-oxireno[1,2]pyrrolo[3,5-b][1,3]oxazin-3-yl]-3-hydroxybutanoate
Molekylvikt (g/mol) 630.7
Formel vikt 630.2741