Learn More
Allyl O-Cumenyl Ether, TRC
Product Code. 30319601
Change view
Click to view available options
Kvantitet:
250 mg
Product Code. Kvantitet
30319601 250 mg
1 options
Product Code. 30319601 Supplier Toronto Research Chemicals Supplier No. TRCA172055250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material Allyl O-Cumenyl Ether
CAS 942-58-5
Molekylformel C12H16O
Kvantitet 250 mg
Synonym 1-(1-Methylethyl)-2-(2-propenyloxy)benzene, 1-(1-Methylethyl)-2-(2-propen-1-yloxy)benzene
InChI formel InChI=1S/C12H16O/c1-4-9-13-12-8-6-5-7-11(12)10(2)3/h4-8,10H,1,9H2,2-3H3
LEDER C=CCOC1=C(C(C)C)C=CC=C1
IUPAC-namn 1-(allyloxy)-2-isopropylbenzene
Molekylvikt (g/mol) 176.26
Formel vikt 176.12