Läs mer
Allylhydrazine Dihydrochloride, TRC
Produktkod. 30324033
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30324033 500 mg
1 options
Produktkod. 30324033 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA544508500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Allylhydrazine Dihydrochloride
CAS 70629-62-8
Molekylformel C3H8N2 . 2HCl
Kvantitet 500 mg
Synonym (Prop-2-en-1-yl)hydrazine dihydrochloride, Hydrazine, 2-propenyl-, dihydrochloride (9CI)
InChI formel InChI=1S/C3H8N2.2ClH/c1-2-3-5-4;;/h2,5H,1,3-4H2;2*1H
LEDER Cl.Cl.NNCC=C
IUPAC-namn prop-2-enylhydrazine;dihydrochloride
Molekylvikt (g/mol) 145.03
Formel vikt 144.0221