missing translation for 'onlineSavingsMsg'
Learn More

Allylpalladium chloride dimer, 98%, Thermo Scientific Chemicals

Catalog Number p-52263
Click to view available options
Quantity:
500 mg
5 g
Packaging:
Glass bottle
12012-95-2
C6H10Cl2Pd2
365.89
MFCD00044874
PENAXHPKEVTBLF-UHFFFAOYSA-L
allylpalladium chloride dimer, bis allyl dichlorodipalladium, diallyldipalladium dichloride, palladium allyl chloride dimer, diallydichlorodipalladium, pi-allyl palladium chloride, allylpalladium ii chloride, bis chloro-pi-allylpalladium, allyl palladium chloride dimer, bis pi-allyl chloropalladium
61538
chloropalladium(1+);prop-1-ene
Cl[Pd].Cl[Pd].c:c:c.c:c:c
12012-95-2
C6H10Cl2Pd2
365.89
MFCD00044874
PENAXHPKEVTBLF-UHFFFAOYSA-L
allylpalladium chloride dimer, bis allyl dichlorodipalladium, diallyldipalladium dichloride, palladium allyl chloride dimer, diallydichlorodipalladium, pi-allyl palladium chloride, allylpalladium ii chloride, bis chloro-pi-allylpalladium, allyl palladium chloride dimer, bis pi-allyl chloropalladium
61538
chloropalladium(1+);prop-1-ene
Cl[Pd].Cl[Pd].c:c:c.c:c:c

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Green-Yellow to Yellow
Assay Percent Range 56.7% min. (Pd)
Packaging Glass bottle
Quantity 500 mg
Fieser 02,109; 05,189; 07,5; 09,132
Solubility Information Solubility in water: decomposes
Formula Weight 365.84
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Allylpalladium chloride dimer, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Allylpalladium chloride dimer
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'