Läs mer
Almotriptan Hydrochloride, TRC
Produktkod. 30327889
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30327889 5 mg
1 options
Produktkod. 30327889 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA5752015MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Almotriptan Hydrochloride
CAS 154323-46-3
Molekylformel C17H26ClN3O2S
Kvantitet 5 mg
Synonym 1H-Indole-3-ethanamine, N,N-dimethyl-5-[(1-pyrrolidinylsulfonyl)methyl]-, hydrochloride (1:1), Pyrrolidine, 1-[[[3-[2-(dimethylamino)ethyl]-1H-indol-5-yl]methyl]sulfonyl]-, monohydrochloride (9CI), Almotriptan hydrochloride
InChI formel InChI=1S/C17H25N3O2S.ClH/c1-19(2)10-7-15-12-18-17-6-5-14(11-16(15)17)13-23(21,22)20-8-3-4-9-20;/h5-6,11-12,18H,3-4,7-10,13H2,1-2H3;1H
LEDER Cl.CN(C)CCc1c[nH]c2ccc(CS(=O)(=O)N3CCCC3)cc12
IUPAC-namn N,N-dimethyl-2-[5-(pyrrolidin-1-ylsulfonylmethyl)-1H-indol-3-yl]ethanamine;hydrochloride
Molekylvikt (g/mol) 371.93