Läs mer
Aloin (mixture of A and B), TRC
Produktkod. 30315799
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30315799 5 g
1 options
Produktkod. 30315799 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA5754235G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Aloin (mixture of A and B)
CAS 8015-61-0
Molekylformel C21H22O9
Kvantitet 5 g
Synonym Aloin (Mixture Of A And B)
InChI formel InChI=1S/C21H22O9/c22-6-8-4-10-14(21-20(29)19(28)17(26)13(7-23)30-21)9-2-1-3-11(24)15(9)18(27)16(10)12(25)5-8/h1-5,13-14,17,19-26,28-29H,6-7H2/t13-,14+,17-,19+,20-,21+/m1/s1
LEDER OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)C2c3cccc(O)c3C(=O)c4c(O)cc(CO)cc24
IUPAC-namn (10S)-1,8-dihydroxy-3-(hydroxymethyl)-10-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-10H-anthracen-9-one
Molekylvikt (g/mol) 418.39
Formel vikt 418.1264