missing translation for 'onlineSavingsMsg'
Läs mer

Aloin (mixture of A and B), TRC

Produktkod. 30320778
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30320778 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30320778 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA5754231G

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material Aloin (mixture of A and B)
CAS 8015-61-0
Molekylformel C21H22O9
Kvantitet 1 g
Synonym Aloin (Mixture Of A And B)
InChI formel InChI=1S/C21H22O9/c22-6-8-4-10-14(21-20(29)19(28)17(26)13(7-23)30-21)9-2-1-3-11(24)15(9)18(27)16(10)12(25)5-8/h1-5,13-14,17,19-26,28-29H,6-7H2/t13-,14+,17-,19+,20-,21+/m1/s1
LEDER OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)C2c3cccc(O)c3C(=O)c4c(O)cc(CO)cc24
IUPAC-namn (10S)-1,8-dihydroxy-3-(hydroxymethyl)-10-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-10H-anthracen-9-one
Molekylvikt (g/mol) 418.39
Formel vikt 418.1264
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.