Läs mer
Alosetron, MedChemExpress
Beskrivning
Alosetron (GR 68755) is a potent and highly selective serotonin 5-HT3 receptor antagonist. Alosetron is used for the research of irritable bowel syndrome (IBS). Alosetron blocks the fast 5HT3-mediated depolarisation of guinea-pig myenteric and submucosal neurons, with IC50 at ∼55 nM. Alosetron attenuates the visceral nociceptive effect of rectal distension in conscious or anaesthetised dogs. Anti-inflammatory effects.
Specifikationer
Specifikationer
| Kemiskt namn eller material | Alosetron |
| CAS | 122852-42-0 |
| Anteckningar om renhetsgrad | Research |
| Molekylformel | C17H18N4O |
| Kvantitet | 10 mg |
| Synonym | GR 68755 GR 68755X |
| LEDER | O=C(C(C1=C2C=CC=C1)=C(N2C)CC3)N3CC4=C(NC=N4)C |
| Molekylvikt (g/mol) | 294.35 |
| Formel vikt | 294.35 |
| Kvalitet | Research |
| Visa mer |