Läs mer
Alpha-[[[2-(4-Methoxyphenyl)-1-methylethyl](phenylmethyl)amino]methyl]-3-nitro-4-(phenylmethoxy)benzenemethanol, TRC
Produktkod. 30480420
Change view
Kvantitet:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30480420 250 mg
30468339 1 g
30467851 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30480420 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM265865250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 43229-67-0
Molekylformel C32H34N2O5
Molekylvikt (g/mol) 526.62
Synonym Benzenemethanol, α-[[[2-(4-methoxyphenyl)-1-methylethyl](phenylmethyl)amino]methyl]-3-nitro-4-(phenylmethoxy)-, α-[[[2-(4-Methoxyphenyl)-1-methylethyl](phenylmethyl)amino]methyl]-3-nitro-4-(phenylmethoxy)benzenemethanol, 1-[4-(Phenylmethoxy)-3-nitrophenyl]-2-[phenylmethyl-[2-(4-methoxyphenyl)-1-methylethyl]amino]ethanol, 2-[[2-(4-Methoxyphenyl)-1-methylethyl](phenylmethyl)amino]-1-[3-nitro-4-(phenylmethoxy)phenyl]ethanol
IUPAC-namn 2-[benzyl-[1-(4-methoxyphenyl)propan-2-yl]amino]-1-(3-nitro-4-phenylmethoxyphenyl)ethanol
LEDER COc1ccc(CC(C)N(CC(O)c2ccc(OCc3ccccc3)c(c2)[N+](=O)[O-])Cc4ccccc4)cc1
Specifikationer
Kemiskt namn eller material alpha-(((2-(4-Methoxyphenyl)-1-methylethyl)(phenylmethyl)amino)methyl)-3-nitro-4-(phenylmethoxy)benzenemethanol
Kvantitet 250 mg
InChI formel InChI=1S/C32H34N2O5/c1-24(19-25-13-16-29(38-2)17-14-25)33(21-26-9-5-3-6-10-26)22-31(35)28-15-18-32(30(20-28)34(36)37)39-23-27-11-7-4-8-12-27/h3-18,20,24,31,35H,19,21-23H2,1-2H3
Formel vikt 526.2468