missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha-2-antiplasmin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13228-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the serpin family of serine protease inhibitors. The protein is a major inhibitor of plasmin, which degrades fibrin and various other proteins. Consequently, the proper function of this gene has a major role in regulating the blood clotting pathway. Mutations in this gene result in alpha-2-plasmin inhibitor deficiency, which is characterized by severe hemorrhagic diathesis. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
Alpha-2-antiplasmin | |
Polyclonal | |
Unconjugated | |
SERPINF2 | |
A2AP, AAP, Alpha 2 antiplasmin, Alpha 2 AP, ALPHA 2 PI, Alpha 2 plasmin inhibitor, Alpha-2-antiplasmin, API, PLI, Serpin F2, SERPINF2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18816, 287527, 5345 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P08697, Q61247 | |
Serpinf2 | |
Alpha-2-antiplasmin Fusion Protein Ag4095 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |