missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha-2-antiplasmin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | Alpha-2-antiplasmin |
---|---|
Concentration | 0.21 mg/mL |
Applications | Immunofluorescence, Immunocytochemistry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16881344
|
Proteintech
13228-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16871344
|
Proteintech
13228-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the serpin family of serine protease inhibitors. The protein is a major inhibitor of plasmin, which degrades fibrin and various other proteins. Consequently, the proper function of this gene has a major role in regulating the blood clotting pathway. Mutations in this gene result in alpha-2-plasmin inhibitor deficiency, which is characterized by severe hemorrhagic diathesis. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
Alpha-2-antiplasmin | |
Immunofluorescence, Immunocytochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
P08697, Q61247 | |
18816, 287527, 5345 | |
Alpha-2-antiplasmin Fusion Protein Ag4095 | |
Primary | |
-20°C | |
SERPINF2 |
0.21 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A2AP, AAP, Alpha 2 antiplasmin, Alpha 2 AP, ALPHA 2 PI, Alpha 2 plasmin inhibitor, Alpha-2-antiplasmin, API, PLI, Serpin F2, SERPINF2 | |
Serpinf2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |