Läs mer
Alpha-[(4,6-Dimethoxy-2-pyrimidinyl)oxy]-Beta-methyl-Beta-phenoxy-benzenepropanoic Acid Methyl Ester (Mixture of Diasteriomers), TRC
Produktkod. 30375230
Change view
Kvantitet:
2.5 mg
Produktkod. Kvantitet
30375230 2.5 mg
1 options
Produktkod. 30375230 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4795282.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Alpha-[(4,6-Dimethoxy-2-pyrimidinyl)oxy]-Beta-methyl-Beta-phenoxy-benzenepropanoic Acid Methyl Ester (Mixture of Diasteriomers)
CAS 159308-02-8
Molekylformel C23H24N2O6
Kvantitet 2.5 mg
Synonym Methyl 2-((4,6-Dimethoxypyrimidin-2-yl)oxy)-3-phenoxy-3-phenylbutanoate
InChI formel InChI=1S/C23H24N2O6/c1-23(16-11-7-5-8-12-16,31-17-13-9-6-10-14-17)20(21(26)29-4)30-22-24-18(27-2)15-19(25-22)28-3/h5-15,20H,1-4H3
LEDER CC(OC1=CC=CC=C1)(C(C(OC)=O)OC2=NC(OC)=CC(OC)=N2)C3=CC=CC=C3
IUPAC-namn methyl 2-((4,6-dimethoxypyrimidin-2-yl)oxy)-3-phenoxy-3-phenylbutanoate
Molekylvikt (g/mol) 424.45
Formel vikt 424.16