Läs mer
alpha-(4-Chlorophenyl)-2-ethyl-1,3-dioxolane-2-acetonitrile, TRC
Produktkod. 30358860
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30358860 1 g
30348826 500 mg
30355590 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30358860 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC4222351G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 55474-41-4
Molekylformel C13 H14 Cl N O2
Molekylvikt (g/mol) 251.71
Synonym 1,3-Dioxolane-2-acetonitrile, α-(4-chlorophenyl)-2-ethyl-, α-(4-Chlorophenyl)-2-ethyl-1,3-dioxolane-2-acetonitrile, (Ξ)-(4-Chlorophenyl)(2-ethyl-1,3-dioxolan-2-yl)acetonitrile
IUPAC-namn 2-(4-chlorophenyl)-2-(2-ethyl-1,3-dioxolan-2-yl)acetonitrile
LEDER CCC1(OCCO1)C(C#N)c2ccc(Cl)cc2
Specifikationer
Kemiskt namn eller material alpha-(4-Chlorophenyl)-2-ethyl-1,3-dioxolane-2-acetonitrile
Kvantitet 1 g
InChI formel InChI=1S/C13H14ClNO2/c1-2-13(16-7-8-17-13)12(9-15)10-3-5-11(14)6-4-10/h3-6,12H,2,7-8H2,1H3
Formel vikt 251.0713