Läs mer
alpha-(4’-Hydroxyphenyl)phloroacetophenone, TRC
Produktkod. 30406341
Change view
Kvantitet:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30406341 100 mg
30397157 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30406341 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH949445100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material alpha-(4'-Hydroxyphenyl)phloroacetophenone
CAS 15485-65-1
Molekylformel C14H12O5
Kvantitet 100 mg
Synonym 2,4,6-Trihydroxyphenyl-4'-hydroxybenzyl Ketone, 2-(p-Hydroxyphenyl)phloroacetophenone, 2-(4-Hydroxyphenyl)-1-(2,4,6-trihydroxyphenyl)ethanone
InChI formel InChI=1S/C14H12O5/c15-9-3-1-8(2-4-9)5-11(17)14-12(18)6-10(16)7-13(14)19/h1-4,6-7,15-16,18-19H,5H2
LEDER Oc1ccc(CC(=O)c2c(O)cc(O)cc2O)cc1
IUPAC-namn 2-(4-hydroxyphenyl)-1-(2,4,6-trihydroxyphenyl)ethanone
Molekylvikt (g/mol) 260.24
Formel vikt 260.0685