Läs mer
Alpha-Acetyl Digoxin, TRC
Produktkod. 30327414
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30327414 5 mg
1 options
Produktkod. 30327414 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA1736605MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material alpha-Acetyldigoxin
CAS 5511-98-8
Molekylformel C43 H66 O15
Kvantitet 5 mg
Synonym Card-20(22)-enolide, 3-[(O-3-O-acetyl-2,6-dideoxy-β-D-ribo-hexopyranosyl-(1→4)-O-2,6-dideoxy-β-D-ribo-hexopyranosyl-(1→4)-2,6-dideoxy-β-D-ribo-hexopyranosyl)oxy]-12,14-dihydroxy-, (3β,5β,12β)-, Digoxin, 3'''-acetate (8CI), Digoxin, acetyl- (6CI), (3β,5β,12β)-3-[(O-3-O-Acetyl-2,6-dideoxy-β-D-ribo-hexopyranosyl-(1→4)-O-2,6-dideoxy-β-D-ribo-hexopyranosyl-(1→4)-2,6-dideoxy-β-D-ribo-hexopyranosyl)oxy]-12,14-dihydroxycard-20(22)-enolide, 3'''-Acetyldigoxin, Desglucolanatoside C, Digoxin, α-acetate, Digoxin, α-acetyl-, Lanatilin, α-Acetyldigoxin
InChI formel InChI=1S/C43H66O15/c1-20-38(49)32(55-23(4)44)18-37(52-20)58-40-22(3)54-36(17-31(40)46)57-39-21(2)53-35(16-30(39)45)56-26-9-11-41(5)25(14-26)7-8-28-29(41)15-33(47)42(6)27(10-12-43(28,42)50)24-13-34(48)51-19-24/h13,20-22,25-33,35-40,45-47,49-50H,7-12,14-19H2,1-6H3/t20-,21-,22-,25-,26+,27-,28-,29+,30+,31+,32+,33-,35+,36+,37+,38-,39-,40-,41+,42+,43+/m1/s1
LEDER C[C@H]1O[C@H](C[C@H](OC(=O)C)[C@@H]1O)O[C@H]2[C@@H](O)C[C@H](O[C@H]3[C@@H](O)C[C@H](O[C@H]4CC[C@@]5(C)[C@H](CC[C@@H]6[C@@H]5C[C@@H](O)[C@]7(C)[C@H](CC[C@]67O)C8=CC(=O)OC8)C4)O[C@@H]3C)O[C@@H]2C
IUPAC-namn [(2R,3R,4S,6S)-6-[(2R,3S,4S,6S)-6-[(2R,3S,4S,6R)-6-[[(3S,5R,8R,9S,10S,12R,13S,14S,17R)-12,14-dihydroxy-10,13-dimethyl-17-(5-oxo-2H-furan-3-yl)-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-3-yl]oxy]-4-hydroxy-2-methyloxan-3-yl]oxy-4-hydroxy-2-methyloxan-3-yl]oxy-3-hydroxy-2-methyloxan-4-yl] acetate
Molekylvikt (g/mol) 822.98
Formel vikt 822.4402