Läs mer
Alpha,Alpha-Dimethyltryptamine, TRC
Produktkod. 30375789
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30375789 50 mg
1 options
Produktkod. 30375789 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD53030550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material alpha,alpha-Dimethyltryptamine
CAS 304-53-0
Molekylformel C12H16N2
Kvantitet 50 mg
Synonym alpha,alpha-Dimethyl-1H-indole-3-ethanamine, 1-(1H-Indol-3-yl)-2-methylpropan-2-amine, 3-(2-Amino-2,2-dimethylethyl)indole, alpha,alpha-Dimethyl-3-indoleethanamine, Ro 3-1638
InChI formel InChI=1S/C12H16N2/c1-12(2,13)7-9-8-14-11-6-4-3-5-10(9)11/h3-6,8,14H,7,13H2,1-2H3
LEDER CC(C)(N)Cc1c[nH]c2ccccc12
IUPAC-namn 1-(1H-indol-3-yl)-2-methylpropan-2-amine
Molekylvikt (g/mol) 188.27
Formel vikt 188.1313