Läs mer
Alpha,Alpha’-Paraxylyl Bismethanethiosulfonate, TRC
Produktkod. 30408884
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30408884 5 mg
1 options
Produktkod. 30408884 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP1932505MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material alpha,alpha'-Paraxylyl Bismethanethiosulfonate
CAS 3948-46-7
Molekylformel C10 H14 O4 S4
Kvantitet 5 mg
Synonym A,A'-Paraxylyl Bismethanethiosulfonate
InChI formel InChI=1S/C10H14O4S4/c1-17(11,12)15-7-9-3-5-10(6-4-9)8-16-18(2,13)14/h3-6H,7-8H2,1-2H3
LEDER CS(=O)(=O)SCc1ccc(CSS(=O)(=O)C)cc1
IUPAC-namn 1,4-bis(methylsulfonylsulfanylmethyl)benzene
Molekylvikt (g/mol) 326.48
Formel vikt 325.9775