Läs mer
alpha-Apo-oxytetracycline, TRC
Produktkod. 30347465
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30347465 10 mg
1 options
Produktkod. 30347465 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA72903510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Alpha-apo-oxytetracycline
CAS 18695-01-7
Molekylformel C22 H22 N2 O8
Kvantitet 10 mg
Synonym (3S,4S,5S)-4-[(1R)-4,5-dihydroxy-9-methyl-3-oxo-1,3-dihydronaphtho[2,3-c]furan-1-yl]-3-(dimethylamino)-2,5-dihydroxy-6-oxocyclohex-1-enecarboxamide, Alpha-apo-oxytetracycline, Oxytetracycline Hydrochloride Imp. D (EP)
InChI formel InChI=1S/C22H22N2O8/c1-7-8-5-4-6-9(25)11(8)16(26)12-10(7)20(32-22(12)31)13-15(24(2)3)17(27)14(21(23)30)19(29)18(13)28/h4-6,13,15,18,20,25-28H,1-3H3,(H2,23,30)/t13-,15-,18-,20-/m0/s1
LEDER CN(C)[C@H]1[C@@H]([C@H](O)C(=O)C(=C1O)C(=O)N)[C@H]2OC(=O)c3c(O)c4c(O)cccc4c(C)c23
IUPAC-namn (3S,4S,5S)-4-[(1R)-4,5-dihydroxy-9-methyl-3-oxo-1H-benzo[f][2]benzofuran-1-yl]-3-(dimethylamino)-2,5-dihydroxy-6-oxocyclohexene-1-carboxamide
Molekylvikt (g/mol) 442.42
Formel vikt 442.1376