Läs mer
(alpha)-Apo-oxytetracycline, TRC
Produktkod. 30338895
Change view
Kvantitet:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30338895 2.5 mg
30338366 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30338895 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA7290352.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Alpha-apo-oxytetracycline
CAS 18695-01-7
Molekylformel C22 H22 N2 O8
Kvantitet 2.5 mg
Synonym (3S,4S,5S)-4-[(1R)-4,5-dihydroxy-9-methyl-3-oxo-1,3-dihydronaphtho[2,3-c]furan-1-yl]-3-(dimethylamino)-2,5-dihydroxy-6-oxocyclohex-1-enecarboxamide, Alpha-apo-oxytetracycline, Oxytetracycline Hydrochloride Imp. D (EP)
InChI formel InChI=1S/C22H22N2O8/c1-7-8-5-4-6-9(25)11(8)16(26)12-10(7)20(32-22(12)31)13-15(24(2)3)17(27)14(21(23)30)19(29)18(13)28/h4-6,13,15,18,20,25-28H,1-3H3,(H2,23,30)/t13-,15-,18-,20-/m0/s1
LEDER CN(C)[C@H]1[C@@H]([C@H](O)C(=O)C(=C1O)C(=O)N)[C@H]2OC(=O)c3c(O)c4c(O)cccc4c(C)c23
IUPAC-namn (3S,4S,5S)-4-[(1R)-4,5-dihydroxy-9-methyl-3-oxo-1H-benzo[f][2]benzofuran-1-yl]-3-(dimethylamino)-2,5-dihydroxy-6-oxocyclohexene-1-carboxamide
Molekylvikt (g/mol) 442.42
Formel vikt 442.1376