missing translation for 'onlineSavingsMsg'
Läs mer

alpha-Bromovalerophenone, TRC

Produktkod. 30330442
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30330442 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30330442 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB689020100MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material alpha-Bromovalerophenone
CAS 49851-31-2
Molekylformel C11 H13 Br O
Kvantitet 100 mg
Synonym 2-Bromo-1-phenylpentan-1-one, 2-Bromovalerophenone
InChI formel InChI=1S/C11H13BrO/c1-2-6-10(12)11(13)9-7-4-3-5-8-9/h3-5,7-8,10H,2,6H2,1H3
LEDER CCCC(Br)C(=O)c1ccccc1
IUPAC-namn 2-bromo-1-phenylpentan-1-one
Molekylvikt (g/mol) 241.12
Formel vikt 240.015
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.